Gibberellic acid
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid
22278.01
22278.02
22278.03
SERA22278.01EA
0
EUR
SERA22278.01
SERA22278.02
SERA22278.03
Gibberellic acid
Gibberellic acid
Formule:
C₁₉H₂₂O₆ Smeltpunt: 234 °C Dichtheid: 1,386 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00079329 CAS nummer: 77-06-5 EINECS: 201-001-0 Merck Index: 13,04430 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...