Lecithin from egg yolk
Leverancier: Serva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
27608.01 27608.02 27608.03
SERA27608.01EA 42 EUR
SERA27608.01 SERA27608.02 SERA27608.03
Lecithin from egg yolk
Lecithin from egg yolk
MDL Nummer: MFCD00131449
CAS nummer: 93685-90-6
EINECS: 297-639-2