Lecithin
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9812.1 9812.2
ROTH9812.1EA 0 EUR
ROTH9812.1 ROTH9812.2
Lecithin
Lecithin
Dichtheid: 1.0305 g/cm³ (20 °C)
MDL Nummer: MFCD00147406
CAS nummer: 8002-43-5
EINECS: 232-307-2
Merck Index: 12,05452