VWR®, Hafnium standard solution, 10,000 mg/L Hf in dil. nitric acid with hydrofluoric acid (max. 1%) (from HfO2) ARISTAR® standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455452BEA 455 EUR
455452B 455454D
VWR®, Hafnium standard solution, 10,000 mg/L Hf in dil. nitric acid with hydrofluoric acid (max. 1%) (from HfO2) ARISTAR® standard for ICP
Standaarden Kationische standaarden Hafnium standaarden
HfO₂ in HNO₃ (2 to 5%) with traces HF.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.