Measuring Cables, CYK10
Leverancier: Endress+Hauser
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CYK10-A031 CYK10-A051 CYK10-A101
ENDRCYK10-A031EA 150 EUR
ENDRCYK10-A031 ENDRCYK10-A051 ENDRCYK10-A101
Measuring Cables, CYK10
Kabels
CYK10 measuring cable is used with all sensors with Memosens plug-in head. Forget about moisture and benefit from completely safe signal transmission.
  • Memosens data cable for all sensors with Memosens plug-in head
  • Maximum process safety via non-contact, inductive and digital signal transmission
  • No contact corrosion, absolutely watertight, no interference potential
  • Increased availability of the measuring point: Digital data transmission provides an automatic error message if the signal is lost
CYK10 guarantees absolute water-proof connection of Memosens sensors to the transmitter. Its inductive coupling shows no contact corrosion and guarantees galvanic isolation of transmitter and medium. Simply forget about limited cable length, interfering potentials or moisture and benefit from completely safe and reliable signal transmission.