4-Hydroxy-4'-iodobiphenyl 98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
OR912760-5G OR912760-25G OR912760-100G OR912760-1G
APOSOR912760-5GEA 90.6 EUR
APOSOR912760-5G APOSOR912760-25G APOSOR912760-100G APOSOR912760-1G
4-Hydroxy-4'-iodobiphenyl 98%
4-Hydroxy-4'-iodobiphenyl
Formule: C₁₂H₉IO
Smeltpunt: 196 - 202 °C
MDL Nummer: MFCD02258843
CAS nummer: 29558-78-9