1-[(4-methoxyphenyl)methyl]pyrazolo[3,4-b]pyridin-3-amine
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
OR55069-100MG OR55069-250MG OR55069-1G
APOSOR55069-100MGEA 869 EUR
APOSOR55069-100MG APOSOR55069-250MG APOSOR55069-1G
1-[(4-methoxyphenyl)methyl]pyrazolo[3,4-b]pyridin-3-amine
1-[(4-methoxyphenyl)methyl]pyrazolo[3,4-b]pyridin-3-amine
Formule: C₁₄H₁₄N₄O
MDL Nummer: MFCD30723358
CAS nummer: 1968547-14-9