1-(3-Bromo-4-hydroxyphenyl)ethanone 97%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
OR019102-1G OR019102-5G
APOSOR019102-1GEA 61.4 EUR
APOSOR019102-1G APOSOR019102-5G
1-(3-Bromo-4-hydroxyphenyl)ethanone 97%
1-(3-Bromo-4-hydroxyphenyl)ethanone
Formule: C₈H₇BrO₂
Smeltpunt: 113 °C
MDL Nummer: MFCD03208974
CAS nummer: 1836-06-2