2-Chloronicotinamide 97%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
OR1214-1G OR1214-5G OR1214-25G
APOSOR1214-1GEA 22 EUR
APOSOR1214-1G APOSOR1214-5G APOSOR1214-25G
2-Chloronicotinamide 97%
2-Chloronicotinamide
Formule: C₆H₅ClN₂O
Smeltpunt: 164 - 167 °C
MDL Nummer: MFCD00006237
CAS nummer: 10366-35-5