2-Cyclopropoxyethyl 4-methylbenzenesulphonate 95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Toluene-4-sulphonic acid 2-cyclopropoxy-ethyl ester, 2-Cyclopropyloxyethyl 4-methylbenzenesulphonate
OR944284-250MG OR944284-1G
APOSOR944284-250MGEA 43.8 EUR
APOSOR944284-250MG APOSOR944284-1G
2-Cyclopropoxyethyl 4-methylbenzenesulphonate 95%
2-Cyclopropoxyethyl 4-methylbenzenesulphonate
Formule: C₁₂H₁₆O₄S
Smeltpunt: 48 °C
MDL Nummer: MFCD22209886
CAS nummer: 862728-59-4