3-Methyl-4-nitrobenzyl bromide 98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
OR927531-5G OR927531-25G OR927531-100G
APOSOR927531-5GEA 95 EUR
APOSOR927531-5G APOSOR927531-25G APOSOR927531-100G
3-Methyl-4-nitrobenzyl bromide 98%
3-Methyl-4-nitrobenzyl bromide
Formule: C₈H₈BrNO₂
Smeltpunt: 46…48 °C
MDL Nummer: MFCD04039920
CAS nummer: 141281-38-1
UN: 3261
ADR: 8,II