3-Nitro-4-(trifluoromethoxy)benzenesulphonyl chloride 95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 3-Nitro-4-(trifluoromethoxy)benzenesulphonyl chloride
PC501730-250MG PC501730-1G
APOSPC501730-250MGEA 263 EUR
APOSPC501730-250MG APOSPC501730-1G
3-Nitro-4-(trifluoromethoxy)benzenesulphonyl chloride 95%
3-Nitro-4-(trifluoromethoxy)benzenesulphonyl chloride
Formule: C₇H₃ClF₃NO₅S
MW: 305.62 g/mol
MDL Nummer: MFCD28801862
CAS nummer: 1351094-07-9
UN: 3265
ADR: 8 III