2-Bromo-5-fluoro-3-(trifluoromethyl)phenol 95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
PC501305-250MG PC501305-1G PC501305-5G
APOSPC501305-250MGEA 210 EUR
APOSPC501305-250MG APOSPC501305-1G APOSPC501305-5G
2-Bromo-5-fluoro-3-(trifluoromethyl)phenol 95%
2-Bromo-5-fluoro-3-(trifluoromethyl)phenol
Formule: C₇H₃BrF₄O
MDL Nummer: MFCD28348577
CAS nummer: 1807145-12-5
UN: 1760
ADR: 8,III