2,4,6-Tris[bis(methoxymethyl)amino]-1,3,5-triazine 98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 2-N,2-N,4-N,4-N,6-N,6-N-hexakis(methoxymethyl)-1,3,5-triazine-2,4,6-triamine, Hexakis(methoxymethyl)melamine, N,N,N',N',N'',N''-Hexakis(methoxymethyl)melamine
OR923794-1G OR923794-5G OR923794-25G
APOSOR923794-1GEA 113 EUR
APOSOR923794-1G APOSOR923794-5G APOSOR923794-25G
2,4,6-Tris[bis(methoxymethyl)amino]-1,3,5-triazine 98%
2,4,6-Tris[bis(methoxymethyl)amino]-1,3,5-triazine
Formule: C₁₅H₃₀N₆O₆
Smeltpunt: 49 °C
Dichtheid: 1,219
MDL Nummer: MFCD00047342
CAS nummer: 3089-11-0