Adamantane-1,3-diamine dihydrochloride 97%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
OR950781-250MG OR950781-1G OR950781-5G
APOSOR950781-250MGEA 145 EUR
APOSOR950781-250MG APOSOR950781-1G APOSOR950781-5G
Adamantane-1,3-diamine dihydrochloride 97%
Adamantane-1,3-diamine dihydrochloride
Formule: C₁₀H₂₀Cl₂N₂
Smeltpunt: 308…350 °C
MDL Nummer: MFCD00137394
CAS nummer: 26562-81-2