meso-2,3-Dimercaptosuccinic acid 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (R,S)-2,3-Dimercaptobutanedioic acid, meso-2,3-Dimercaptobutanedioic acid, Succimer, (R,S)-2,3-Dimercaptosuccinic acid, (2R,3S)-rel-2,3-Dimercaptosuccinic acid

226670050 226670250 226671000
ACRO226670050EA 85.1 EUR
ACRO226670050 ACRO226670250 ACRO226671000
meso-2,3-Dimercaptosuccinic acid 98%
meso-2,3-Dimercaptosuccinic acid
Formule: C₄H₆O₄S₂
MW: 182.22 g/mol
Smeltpunt: 196…198 °C (ontbinding)
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00064799
CAS nummer: 304-55-2
EINECS: 206-155-2
Merck Index: 12,09034

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤1 % (1 g, 105°C, 3 hrs)
Sulfated ash ≤0.2 %
Titration with NaOH ≥97.5 %
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR