4-Bromo-m-toluic acid
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 4-Bromo-3-methylbenzoic acid, 2-Bromo-5-carboxytoluene, 4-Bromo-m-toluic acid
OR3056-250MG OR3056-1G
APOSOR3056-250MGEA 22 EUR
APOSOR3056-250MG APOSOR3056-1G
4-Bromo-m-toluic acid
4-Bromo-m-toluic acid
Formule: C₈H₇BrO₂
Smeltpunt: 212 - 216 °C
MDL Nummer: MFCD00039526
CAS nummer: 7697-28-1