Pyridine-2-carbaldehyde 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

OR5051-25G OR5051-500G
APOSOR5051-25GEA 0 EUR
APOSOR5051-25G APOSOR5051-500G
Pyridine-2-carbaldehyde 99%
Pyridine-2-carbaldehyde
Formule: C₆H₅NO
Kookpunt: 179 - 181 °C
Smeltpunt: –21 °C
Dichtheid: 1,126
Vlampunt: 73 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006290
CAS nummer: 1121-60-4
UN: 2810
ADR: 6.1,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR