2,2,3,3,3-Pentafluoropropyl acrylate 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
PC5785-5G
PC5785-1G
APOSPC5785-5GEA
117
EUR
APOSPC5785-5G
APOSPC5785-1G
2,2,3,3,3-Pentafluoropropyl acrylate 97%
2,2,3,3,3-Pentafluoropropyl acrylate
Formule:
C₆H₅F₅O₂ Kookpunt: 50/100 mm °C Dichtheid: 1,32 Vlampunt: 23 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00039257 CAS nummer: 356-86-5 UN: 3272 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...