4-Bromo-3-fluoro-2-methoxybenzonitrile 98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
PC500667-250MG PC500667-1G
APOSPC500667-250MGEA 186 EUR
APOSPC500667-250MG APOSPC500667-1G
4-Bromo-3-fluoro-2-methoxybenzonitrile 98%
4-Bromo-3-fluoro-2-methoxybenzonitrile
Formule: C₈H₅BrFNO
MW: 230.0338 g/mol
MDL Nummer: MFCD23707400
CAS nummer: 1427452-84-3