Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)]palladium(II) 98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: P(t-Bu)3 Pd G2
OR019112-1G OR019112-5G
APOSOR019112-1GEA 58.4 EUR
APOSOR019112-1G APOSOR019112-5G
Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)]palladium(II) 98%
Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)]palladium(II)
Formule: C₂₄H₃₇ClNPPd
MW: 512.41 g/mol
Smeltpunt: 167…170 °C
MDL Nummer: MFCD21608496
CAS nummer: 1375325-71-5