2,6-Difluoro-3-(trifluoromethoxy)benzonitrile 95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
PC502468-250MG PC502468-1G PC502468-5G
APOSPC502468-250MGEA 176 EUR
APOSPC502468-250MG APOSPC502468-1G APOSPC502468-5G
2,6-Difluoro-3-(trifluoromethoxy)benzonitrile 95%
2,6-Difluoro-3-(trifluoromethoxy)benzonitrile
Formule: C₈H₂F₅NO
MDL Nummer: MFCD28791008
CAS nummer: 1806290-43-6
UN: 2810
ADR: 6.1,III