3-(2,4,5-Trichlorophenyl)-1H-pyrazole
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
OR64746-100MG OR64746-250MG
APOSOR64746-100MGEA 110 EUR
APOSOR64746-100MG APOSOR64746-250MG
3-(2,4,5-Trichlorophenyl)-1H-pyrazole
3-(2,4,5-Trichlorophenyl)-1H-pyrazole
Formule: C₉H₅Cl₃N₂
MDL Nummer: MFCD32862646
CAS nummer: 2645418-49-9