3-(Aminomethyl)-1-(cyclopropyl)-5-(trifluoromethyl)-1H-pyrazole dihydrochloride 95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
PC512032-250MG PC512032-1G PC512032-5G
APOSPC512032-250MGEA 162 EUR
APOSPC512032-250MG APOSPC512032-1G APOSPC512032-5G
3-(Aminomethyl)-1-(cyclopropyl)-5-(trifluoromethyl)-1H-pyrazole dihydrochloride 95%
3-(Aminomethyl)-1-(cyclopropyl)-5-(trifluoromethyl)-1H-pyrazole dihydrochloride