5-(Difluoromethyl)-2-fluoroaniline 95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
PC55857-250MG PC55857-1G PC55857-5G
APOSPC55857-250MGEA 158 EUR
APOSPC55857-250MG APOSPC55857-1G APOSPC55857-5G
5-(Difluoromethyl)-2-fluoroaniline 95%
5-(Difluoromethyl)-2-fluoroaniline
Formule: C₇H₆F₃N
MDL Nummer: MFCD24645334
CAS nummer: 1374298-68-6