tert-Butyl 4-(2,2-difluoroethyl)piperazine-1-carboxylate 95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
PC57443-250MG PC57443-1G
APOSPC57443-250MGEA 121 EUR
APOSPC57443-250MG APOSPC57443-1G
tert-Butyl 4-(2,2-difluoroethyl)piperazine-1-carboxylate 95%
tert-Butyl 4-(2,2-difluoroethyl)piperazine-1-carboxylate
Formule: C₁₁H₂₀F₂N₂O₂
MDL Nummer: MFCD20441471
CAS nummer: 1225380-87-9