3,5-Difluoro-4-(trifluoromethyl)benzonitrile 95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
PC57489-1G PC57489-5G
APOSPC57489-1GEA 526 EUR
APOSPC57489-1G APOSPC57489-5G
3,5-Difluoro-4-(trifluoromethyl)benzonitrile 95%
3,5-Difluoro-4-(trifluoromethyl)benzonitrile
Formule: C₈H₂F₅N
MDL Nummer: MFCD01631623
CAS nummer: 261945-30-6
UN: 3439
ADR: 6.1,III