2-Bromo-5-chloro-3-fluorobenzoic acid
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
PC57497-1G PC57497-5G
APOSPC57497-1GEA 957 EUR
APOSPC57497-1G APOSPC57497-5G
2-Bromo-5-chloro-3-fluorobenzoic acid
2-Bromo-5-chloro-3-fluorobenzoic acid
Formule: C₇H₃BrClFO₂
MDL Nummer: MFCD24119836
CAS nummer: 1529288-48-9