4,4'-Bipyridine -1,1'-dioxide 98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
OR22806-1G OR22806-5G OR22806-10G
APOSOR22806-1GEA 76 EUR
APOSOR22806-1G APOSOR22806-5G APOSOR22806-10G
4,4'-Bipyridine -1,1'-dioxide 98%
4,4'-Bipyridine -1,1'-dioxide
Formule: C₁₀H₈N₂O₂
Smeltpunt: 298 - 306 °C
MDL Nummer: MFCD00134383
CAS nummer: 24573-15-7