4-Bromo-3-fluoro-5-methylaniline 98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
PC908562-250MG PC908562-1G PC908562-5G
APOSPC908562-250MGEA 64.3 EUR
APOSPC908562-250MG APOSPC908562-1G APOSPC908562-5G
4-Bromo-3-fluoro-5-methylaniline 98%
4-Bromo-3-fluoro-5-methylaniline
Formule: C₇H₇BrFN
MDL Nummer: MFCD19686156
CAS nummer: 1356113-07-9