1-Adamantanethiol 98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
OR309163-5G OR309163-25G
APOSOR309163-5GEA 87.6 EUR
APOSOR309163-5G APOSOR309163-25G
1-Adamantanethiol 98%
1-Adamantanethiol
Formule: C₁₀H₁₆S
Smeltpunt: 103 - 105 °C
MDL Nummer: MFCD00213530
CAS nummer: 34301-54-7
UN: 2811
ADR: 6.1,III