Safety Knife, SECUNORM 185
Leverancier: Martor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MARO185004.17EA 20.2 EUR
MARO185004.17
Safety Knife, SECUNORM 185
Messen
The automatic blade retraction of the SECUNORM 185 guarantees a high degree of safety from cut injuries. It provides maximum safety with ergonomic and ceramic blades.
  • Four cutting edge blade
  • Lanyard hole
  • Tool-less blade change
  • Abrasion resistant
  • For right and left handers
  • Cutting depth: 9 mm
Weight knife 31,8 g
Size single unit box: 103×10×29 mm
Weight single unit box: 34,5 g
Quantity in outer pack: 20 knives
Size outer pack: 118×52×108 mm
Weight outer pack: 725,0 g
Lots of knife requiring little space. With ceramic blade. Efficiency is important when opening cartons and pallets. Just what the nimble and robust SECUNORM 185 has beendesigned for. The compact safety knife impresses with features rarely found in this product group, such as a stainless steel handle, a large slider and easy blade change. changing. Other benefits are the sharp, long-lasting ceramic blade and automatic blade retraction. Robust and practical: The SECUNORM 185's handle is made of stainless steel, designed for frequent cutting and long service. The knife has an ergonomic practical format and lightweight design to make cutting easier.
Optimal slider: The double-sided slider is suitable for both right and left handers. It has a large surface for perfect operation even when wearing gloves. Grooves on the slider improve slip resistance. Safe blade change: Simply hold down the blade change button and pull the slider out from the front to access the blade. The plastic insert and handle remain together so that nothing will be lost.
Practical eyelet: The SECUNORM 185 is equipped with a eyelet for attaching a strap, securing the knife to be on hand as soon as the next package needs opening. Four cutting edge blade: The blade has four cutting edges that can be used one after the other. This makes economic and ecological sense and, with a fresh cutting edge, you can always cut like new.
Main cutting material: Cardboard: up to 2-ply; wrapping, stretch and shrink foil; tape; plastic strapping band; layers of foil or paper; yarn, cord; textile, material.
Certificaten: GS certificate no. 180823-7255-55094-2021.
Verpakking: Packaging unit: 1 in single unit box.