(R)-(-)-Mandelic acid
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid
10007128-25 10007128-50 10007128-100 10007128-250
CAYM10007128-25EA 92.5 EUR
CAYM10007128-25 CAYM10007128-50 CAYM10007128-100 CAYM10007128-250
(R)-(-)-Mandelic acid
(R)-(-)-Mandelic acid
Formule: C₈H₈O₃
MW: 152.15 g/mol
Kookpunt: 322
Smeltpunt: 130…132 °C
Dichtheid: 1.341 g/cm³ (20 °C)
Vlampunt: >190 °C
MDL Nummer: MFCD00064251
CAS nummer: 611-71-2
EINECS: 210-276-6