trans-4-Hydroxy-L(-)-proline
Leverancier: BACHEM BIOCHEMICA
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
4002357.0025
4002357.0100
4002357.0250
BACE4002357.0025EA
62.1
EUR
BACE4002357.0025
BACE4002357.0100
BACE4002357.0250
trans-4-Hydroxy-L(-)-proline
trans-4-Hydroxy-L(-)-proline
Formule:
C₅H₉NO₃ Kookpunt: 296 °C (1013 hPa) Smeltpunt: 206 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00064320 CAS nummer: 51-35-4 EINECS: 200-091-9 Merck Index: 13,04866 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...