Cleanroom Lab Coats, Alsistrong, Light Blue
Leverancier: ALSICO HIGHTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
103964-M 103964-2XL 103964-L 103964-S 103964-XS 103964-XL
ALSO103964-MEA 0 EUR
ALSO103964-M ALSO103964-2XL ALSO103964-L ALSO103964-S ALSO103964-XS ALSO103964-XL
Cleanroom Lab Coats, Alsistrong, Light Blue
Werkkleding
This cleanroom coat is made of Alsistrong® (98% polyester and 2% carbon).