TEGERA® 12910 Chemical and Liquid Protection Gloves
Leverancier: EJENDALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12910-10 12910-11 12910-7 12910-8 12910-9
EJEN12910-10EA 140.94 EUR
EJEN12910-10 EJEN12910-11 EJEN12910-7 EJEN12910-8 EJEN12910-9
TEGERA® 12910 Chemical and Liquid Protection Gloves
Handschoenen
A super-long, high-safety glove 0,3 mm/11,8 mil with sandy finish for a secure grip. The glove protects against chemicals and is perfect to use when cleaning tanks. It is oil resistant thanks to triple PVC coating and stays soft in cold conditions.
  • High level of protection
  • Good fit
  • Durable
  • Good grip
  • Comfortable
  • Waterproof
  • Phthalate-free
  • Oil and grease resistant
Prevents risk of corrosive injuries, puncturing, abrasion injuries, contact with dirt, contact with chemicals, contact with moisture, contact with damp, contact with oil.
Primary environments of use: Chemical risk environments, microbiological risk environments, environments hazardous to health, corrosive environments, wet environments, moist environments, oily and greasy environments, dirty environments.
Primary areas of use: Hvac installation, petro-chemical work, chemical work, chemical technology work, paper industry work, decontamination, sanitation, cleaning, laboratory work, fishing industry work, marine work.
Certificaten: EN ISO 21420:2020, EN 388:2016+A1:2018, 4121X, EN 374-3:2003, EN ISO 374-1:2016/A1:2018 Type B, JKL.