Dicotyledon plant stem
Leverancier: 3B Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
763-0138EA 244 EUR
763-0138
Dicotyledon plant stem
Modellen Plantkunde modellen
Phaseolus vulgaris. Model of dicotyledonous stem (beans) magnified 250X.
Stem cell types are shown longitudinally and in cross section.