Leader, function models
Leverancier: 3B Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1000159 1000161 1000165
763-0086EA 0 EUR
763-0086 763-0087 763-0089
Leader, function models
Modellen Menselijke biologie modellen
Shoulder joint: Life-size plastic moldings of the upper part of the humerus; shoulder blades and collarbones with ligaments. Movements that can be studied: the arm is lifted (abduction), moved forwards and backwards (anteversion or retroversion) and rotation inwards and outwards. On stand.
Hip joint: Natural casting. Consists of the hip bone and the upper part of the femur with ligaments. On stand. Movements that can be shown are anteversion, retroversion and abduction.
Elbow joint: Natural casting. Consists of upper arm and forearm with ligaments. On stand. Functions that can be shown are bending and stretching of the upper and forearm as well as the rotational movements of the arm.