Calibrated Disposable Plastic Inoculation Loops
Leverancier: Copan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8175CS10H 178CS10 178CS20 179CS10 179CS20 8177CS10H 8175CS20H 8177CS20H 176CS20 178CS40 179CSR40 183CS10 8175CSR40H 8177CSR40H
710-0195EA 165 EUR
710-0195 710-0191 710-0192 710-0193 710-0194 710-0197 710-0196 710-0198 COPA176CS20 COPA178CS40 COPA179CSR40 COPA183CS10 COPA8175CSR40H COPA8177CSR40H
Calibrated Disposable Plastic Inoculation Loops
Uitstrijkspatels Entogen en -naalden
COPAN calibrated plastic inoculation loops have an ultra-smooth surface to avoid gouging agar when planting and streaking.
Loops are free of lubricants, oils, and electrostatic charges.