Straight Tubing Connectors
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H195030000 H195100000 H19508-0000 H19509-0000
BELAH195030000EA 129 EUR
BELAH195030000 BELAH195100000 BELAH19508-0000 BELAH19509-0000
Straight Tubing Connectors
Fittingen en connectoren
These straight fittings ensure a tight, secure connection between lengths of tubing having the same inner diameter.
  • Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Verpakking: Pack of 12.