Ethyl cyanoacetate 98+%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
118280010 118282500 118280051
ACRO118280010EA 113 EUR
ACRO118280010 ACRO118282500 ACRO118280051
Ethyl cyanoacetate 98+%
Ethyl cyanoacetate
Formule: C₅H₇NO₂
Kookpunt: 209 °C (1013 hPa)
Smeltpunt: –22 °C
Dichtheid: 1,06 g/cm³
Vlampunt: 110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001940
CAS nummer: 105-56-6
EINECS: 203-309-0
Merck Index: 13,03821
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4190 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥98.0 %
Color scale ≤50 APHA
Free acid ≤10 mg/kg (as cyanoacetic acid)