Furfuryl mercaptan 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Furanmethanethiol, 2-Furanmethanthiol, 2-Furylmethanethiol
119810250 119811000
ACRO119810250EA 43.1 EUR
ACRO119810250 ACRO119811000
Furfuryl mercaptan 97%
Furfuryl mercaptan
Formule: C₅H₆OS
MW: 114.17 g/mol
Kookpunt: 155
Dichtheid: 1.13 g/cm³
Vlampunt: 45 °C
MDL Nummer: MFCD00003254
CAS nummer: 98-02-2
EINECS: 202-628-2
UN: 1228
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5290 to 1.5330 (20°C, 589 nm)
GC ≥96.0 %