Isopreen 98% stabilised
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
122670010 122670025 122671000 122675000
ACRO122670010EA 93.5 EUR
ACRO122670010 ACRO122670025 ACRO122671000 ACRO122675000
Isopreen 98% stabilised
Isopreen
Formule: C₅H₈
Kookpunt: 34 °C (1013 hPa)
Smeltpunt: –146 °C
Dichtheid: 0,68 g/cm³
Vlampunt: –48 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008600
CAS nummer: 78-79-5
EINECS: 201-143-3
UN: 1218
ADR: 3,I
Merck Index: 13,05220
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4210 to 1.4230 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 100 to 250 ppm 4-tert-Butylcatechol