1,6-Bis(dimethylamino)hexane 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: N,N,N',N'-Tetramethylhexamethylenediamine, N,N,N',N'-Tetramethyl-1,6-diaminohexane, N1,N1,N6,N6-Tetramethylhexane-1,6-diamine, N,N,N',N'-Tetramethyl-1,6-hexanediamine
147490010 147490250 147492500
ACRO147490010EA 1119 EUR
ACRO147490010 ACRO147490250 ACRO147492500
1,6-Bis(dimethylamino)hexane 99%
1,6-Bis(dimethylamino)hexane
Formule: C₁₀H₂₄N₂
Kookpunt: 209…210 °C (760 mmHg)
Smeltpunt: –46 °C
Dichtheid: 0,806 g/cm³
Vlampunt: 73 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008339
CAS nummer: 111-18-2
EINECS: 203-842-9
UN: 2922
ADR: 8,I
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4350 to 1.4370 (20°C, 589 nm)
GC ≥98.5 %