Tetramethylammonium fluoride tetrahydrate 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
148070050 148070250
ACRO148070050EA 32.1 EUR
ACRO148070050 ACRO148070250
Tetramethylammonium fluoride tetrahydrate 98%
Tetramethylammonium fluoride tetrahydrate
Formule: C₄H₁₂FN·4H₂O
Smeltpunt: 39…44 °C
Dichtheid: 1,115 g/cm³ (20 °C)
Vlampunt: >112 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149968
CAS nummer: 17787-40-5
EINECS: 206-769-0
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Colorless to white Low melting crystalline mass
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %