Methyl dichloroacetate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

148621000 148625000
ACRO148621000EA 18.3 EUR
ACRO148621000 ACRO148625000
Methyl dichloroacetate 99%
Methyl dichloroacetate
Formule: C₃H₄Cl₂O₂
Kookpunt: 143 °C (1013 hPa)
Smeltpunt: –52 °C
Dichtheid: 1,381 g/cm³
Vlampunt: 80 °C
MDL Nummer: MFCD00000843
CAS nummer: 116-54-1
EINECS: 204-146-8
UN: 2299
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4411 to 1.4431 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR