Methylchlooracetaat 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
148630010
148630025
148630100
ACRO148630010EA
76.2
EUR
ACRO148630010
ACRO148630025
ACRO148630100
Methylchlooracetaat 98%
Methylchlooracetaat
Formule:
C₃H₅ClO₂ Kookpunt: 130…132 °C (760 mmHg) Smeltpunt: –33 °C Dichtheid: 1,23 g/cm³ Vlampunt: 47 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000931 CAS nummer: 96-34-4 EINECS: 202-501-1 UN: 2295 ADR: 6.1, Merck Index: 12,06122 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4205 to 1.4225 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...