2-Tolualdehyde 98% stabilised
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: o-Tolualdehyde, 2-Methylbenzaldehyde
152431000 152432500 152430250
ACRO152431000EA 107 EUR
ACRO152431000 ACRO152432500 ACRO152430250
2-Tolualdehyde 98% stabilised
2-Tolualdehyde
Formule: C₈H₈O
MW: 120.15 g/mol
Kookpunt: 199…200 °C (760 mmHg)
Smeltpunt: –35 °C
Dichtheid: 1.039 g/cm³
Vlampunt: 67 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003338
CAS nummer: 529-20-4
EINECS: 208-452-2
Merck Index: 12,09665
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5460 to 1.5490 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 0.1 to 0.5 % hydroquinone