5-Chlorovaleryl chloride 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 5-Chlorovaleroyl chloride, 5-Chloropentanoyl chloride

154770100 154770250 154771000
ACRO154770100EA 92.6 EUR
ACRO154770100 ACRO154770250 ACRO154771000
5-Chlorovaleryl chloride 96%
5-Chlorovaleryl chloride
Formule: C₅H₈Cl₂O
Kookpunt: 97…99 °C (1 mmHg)
Dichtheid: 1,2 g/cm³
Vlampunt: 90 °C
MDL Nummer: MFCD00000758
CAS nummer: 1575-61-7
EINECS: 216-403-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4630 to 1.4650 (20°C, 589 nm)
GC ≥95 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR