trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Titriplex® IV
155000250 155001000 155000025 155005000
ACRO155000250EA 72 EUR
ACRO155000250 ACRO155001000 ACRO155000025 ACRO155005000
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 98%
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
Formule: C₁₄H₂₂N₂O₈·1H₂O
MW: 364.35 g/mol
Smeltpunt: >210 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00066429
CAS nummer: 125572-95-4
EINECS: 236-308-9
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Complexometric 97.5 to 102.5 %
Loss on drying 4.0 to 6.9 % (1g,150°C,6h)
Sulfated ash ≤0.3 %